Order Entry
Germany
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

Trypsin 0,25% EDTA 0,02% in HBSS, without calcium, without magnesium, with phenol red, 100 mL
Catalog # L0932-100
Supplier:   Biowest
Trypsin
Catalog # L0932-100
Supplier:  Biowest
CAS Number:  
Trypsin
Trypsin

Trypsin 0,25% EDTA 0,02% in HBSS, without calcium, without magnesium, with phenol red, 100 mL

CAS Number:  
Price
25,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Trypsin 0,25% EDTA 0,02% in HBSS, without calcium, without magnesium, with phenol red

Specifications