Order Entry
Germany
Orders LinkContactUsLinkComponent
 

G418 solution

100 mL
Catalog # L0015-100
Supplier:   Biowest
G418 solution
Catalog # L0015-100
Supplier:  Biowest
CAS Number:  49863-47-0
G418 solution
G418 solution

100 mL

CAS Number:  49863-47-0
Price
380,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Antibiotic G418 is an aminoglycoside antibiotic similar in structure to gentamycin. It exhibits toxicity towards both eukaryotic and prokaryotic cells. The optimal concentration for selection and maintenance must be determined for each cell line.

  • Clear to very slightly hazy liquid
  • Shelf life of 24 months
  • Storage temperature of –20 °C

 : This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Synonyms:  G-418, (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol
  • CAS Number:49863-47-0
  • Formula:C₂₀H₄₀N₄O₁₀
  • MDL Number:MFCD30541267
  • Molecular Weight:496.552 g/mol

 

Specifications

Specifications

  • Pk:
    100 mL

Specifications