Order Entry
Germany
Orders LinkContactUsLinkComponent
G418 solution

G418 solution

20 mL
Catalog # L0015-020
Supplier:   Biowest
G418 solution
Catalog # L0015-020
Supplier:  Biowest
CAS Number:  49863-47-0
G418 solution
G418 solution

20 mL

CAS Number:  49863-47-0
Price
85,50 €
Log in for availability
1 * 20 mL
Product Details & Documents

Antibiotic G418 is an aminoglycoside antibiotic similar in structure to gentamycin. It exhibits toxicity towards both eukaryotic and prokaryotic cells. The optimal concentration for selection and maintenance must be determined for each cell line.

  • Clear to very slightly hazy liquid
  • Shelf life of 24 months
  • Storage temperature of –20 °C

Caution: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Synonyms:  G-418, (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol
  • CAS Number:49863-47-0
  • Formula:C₂₀H₄₀N₄O₁₀
  • MDL Number:MFCD30541267
  • Molecular Weight:496.552 g/mol

Documents

Specifications

Specifications

  • Pk:
    20 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.