Order Entry
Germany
Orders LinkContactUsLinkComponent
Glutamine

Glutamine

L-glutamic acid (powder), 100 g
Catalog # P1011-100GR
Supplier:   Biowest
Glutamine
Catalog # P1011-100GR
Supplier:  Biowest
CAS Number:  
Glutamine
Glutamine

L-glutamic acid (powder), 100 g

CAS Number:  
Price
85,90 €
Log in for availability
1 * 100 g
Product Details & Documents

L-glutamine is an essential amino acid required by virtually all mammalian and insect cells grown in culture. It is a crucial component of many cell culture media and serves as a major energy source for cells in culture. L-glutamine is very stable as a dry powder and as a frozen solution. However, in liquid media or stock solutions, L-glutamine can degrade relatively rapidly. L-glutamine is also more labile in cell culture solution than other amino acids.

Dipeptide derivatives of L-glutamine (stable glutamine) prevent the intramolecular cyclisation reaction associated with solutions of L-glutamine. These derivatives are therefore stable in solution and allow the formulation of cell culture media containing L-glutamine that may be stored at 4 °C for extended periods. Solutions containing these derivatives can even be autoclaved without appreciable degradation of the product (30 minutes at 121 °C results in <5% loss of the product).

The dipeptide derivatives are metabolised within the cells to yield L-glutamine plus the second amino acid. This results in more consistent delivery of L-glutamine to cells and avoids toxic build-up of ammonia in cell cultures. This feature can be especially important for ammonia-sensitive cell lines.

Documents

Specifications

Specifications

  • Pk:
    100 g
  • Description:
    L-glutamic acid (powder)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.