Orders LinkContactUsLinkComponent
Microorganism tests, preservation system, CryoInstant
Microorganism tests, preservation system, CryoInstant
CryoInstant, white, 100
Catalog # DELT409113/1
Supplier:   DELTALAB
Microorganism tests, preservation system, CryoInstant
Catalog # DELT409113/1
Supplier:  DELTALAB
CAS Number:  
Microorganism tests, preservation system, CryoInstant
Microorganism tests, preservation system, CryoInstant
CryoInstant, white, 100
CAS Number:  
Price
526,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
CryoInstant is a preservation system for storing microbiological culture consisting on 2 ml cryovials containing 25 porous beads and cryopreservative-added broth.
  • Perfect freezing storage system (Save space in the freezers when storing)
  • Can be used to obtain up to 25 replicates from the original microorganism (to use them during many years)
  • Ease the recovery of the stored strain streaking an inoculated bead directly onto solid media
  • Prevent changes and repetitive thawing/freezing of the rest of beads
  • Avoid the growth of ice when recovering
  • Improve recovery rates of frozen cultures by adding cryopreservative to broth
  • Minimise the risk of cross contamination
The external thread cryovials are made of PP with smooth inner surface which prevents from
contamination. Their caps embody a silicone washer to ensure a positive leakproof seal. Tubes and caps withstand up to –190 ºC.
Packaging: Cryovials are packed in carton boxes with 100 units. Each box is supplied plastic-wrapped and labelled with code reference, batch, expiration date and specifying the colour of the caps and beads.
Specifications
Specifications
  • Pk:
    100
  • Description:
    CryoInstant, white