Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ethylacetat-INFRASOLV Gehalt: >99,5% Abdampfrückstand:<20mg/l Farbzahl:< 10 Hazen 1 * 5 L
Catalog # LBCL5L588099
Supplier:
LABC LABORTECHNIK
Expand image
Ethylacetat-INFRASOLV Gehalt: >99,5% Abdampfrückstand:<20mg/l Farbzahl:< 10 Hazen 1 * 5 L
Catalog # LBCL5L588099
Supplier:
LABC LABORTECHNIK