Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillary, non-swaged (from short column to valve with M4 port), 0,13 m, I.Ø 0,12 mm
Catalog # HEWL5067-4735
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWL5067-4735
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems
Capillary, non-swaged (from short column to valve with M4 port), 0,13 m, I.Ø 0,12 mm
Price
138,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Description:
    Capillary, non-swaged (from short column to valve with M4 port), 0,13 m, I.Ø 0,12 mm
  • Seq:
    75