Capillaries for LC systems
Capillaries, pre-swaged, pump to thermostatted autosampler, 450 mm, I.Ø 0,17 mm
HEWL5067-4658
: Agilent
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Description:Capillaries, pre-swaged, pump to thermostatted autosampler, 450 mm, I.Ø 0,17 mm
- Seq:58