Order Entry
Germany
Orders LinkContactUsLinkComponent
Capillary columns, DB-624

Capillary columns, DB-624

7890/6890 LTM module, Capillary, DB-624, 1,00 µm, 0,18 mm, 20 m
Catalog # HEWL121-1324LTM
Supplier:   Agilent
Capillary columns, DB-624
Catalog # HEWL121-1324LTM
Supplier:  Agilent
Capillary columns, DB-624
Capillary columns, DB-624

7890/6890 LTM module, Capillary, DB-624, 1,00 µm, 0,18 mm, 20 m

Price
1 900,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These columns are recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS volatiles analysis. They are specifically designed for the analysis of volatile priority pollutants and residual solvents.

  • Excellent inertness for active compounds
  • Bonded and cross-linked
  • Solvent rinsable
Specifications
  • Column Format:
    Capillary
  • Film thickness:
    1,00 µm
  • Ø int.:
    0,18 mm
  • Description:
    7890/6890 LTM module
  • Length:
    20 m
  • Phase:
    DB-624
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.