Universal vial caps
Universal vial caps, PA 800 plus or PA 800 enhanced instruments, Blue
Supplier: EKSIGENT
Jump to:
Product Details & Documents
Universal caps for use with vials.
Universal vial caps are designed for use only with CESI 8000, PA 800 Enhanced or PA 800 plus instruments and should not be reused.
Documents
Certificates
Search by Lot NumberSpecifications
- For:PA 800 plus or PA 800 enhanced instruments
- Description:Universal vial caps
- Cap colour:Blue