Standard solution cyanide 100 mg CN-/l KCN in sodium hydroxide solution 0.4 mol/l DIN 38405-7:2002, DEV D7 DIN 38405-13:2011, DEV D1 3-1 1 * 100 mL
Catalog # KRAF10503.0700
Lieferant: Bernd Kraft
Standard solution cyanide 100 mg CN-/l KCN in sodium hydroxide solution 0.4 mol/l DIN 38405-7:2002, DEV D7 DIN 38405-13:2011, DEV D1 3-1 1 * 100 mL
Catalog # KRAF10503.0700
Lieferant: Bernd Kraft
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Lieferant: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Lieferant: Merck
Ethanol 99.5 % denatured with 1 % MEK 99,5% ≥99.5%, GPR RECTAPUR® (denatured with 1% MEK)
Lieferant: VWR Chemicals
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
Lieferant: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Lieferant: Merck
VWR® Aquaresist Water Bath Protective Agent
Lieferant: VWR Chemicals
Acetone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Lieferant: Dr. Weigert
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Lieferant: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Lieferant: VWR Chemicals
Chromium(VI) standard solution 1.000 g Cr6+/l K2CrO4 in water Content: 0,5 l 1 * 500 mL
Lieferant: Bernd Kraft