Respiratory filters
Combined filter, 92, A2B2E2K1 P2
Catalog # 111-1558
Supplier: M S A
Jump to:
Product Details & Documents
Screw-in gas or combination filter for protection against toxic gases, vapours or dusts.
: EN 148-1
Specifications
- Type:Combined filter, 92
- Seq:04
- Class:A2B2E2K1 P2