ContactUsLinkComponent
Ultra Phenyl x 1.8µm particle size x 120Å pore size, 50mm length x 2.1mm ID, Ultrahigh pressure hardware ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2UPH-18L02U
Ultra Phenyl x 1.8µm particle size x 120Å pore size, 50mm length x 2.1mm ID, Ultrahigh pressure hardware ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2UPH-18L02U
Frequently Bought Together