ContactUsLinkComponent
C18G x 5µm particle size x 300Å pore size, 250mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-05R02P
C18G x 5µm particle size x 300Å pore size, 250mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-05R02P
Frequently Bought Together