ContactUsLinkComponent
Silica x 3µm particle size x 120Å pore size, 150mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2SIL-03N46S
Silica x 3µm particle size x 120Å pore size, 150mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2SIL-03N46S
Frequently Bought Together