ContactUsLinkComponent
C18Q x 5µm particle size x 100Å pore size, 250mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-05R03P
C18Q x 5µm particle size x 100Å pore size, 250mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-05R03P
Frequently Bought Together