ContactUsLinkComponent
C18Q x 5µm particle size x 100Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C183-05M15K
C18Q x 5µm particle size x 100Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C183-05M15K
Frequently Bought Together