ContactUsLinkComponent
SCX x 5µm particle size x 120Å pore size, 250mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2SCX-05R03S
SCX x 5µm particle size x 120Å pore size, 250mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2SCX-05R03S
Frequently Bought Together