ContactUsLinkComponent
C18Q x 5µm particle size x 100Å pore size, 50mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-05L46S
C18Q x 5µm particle size x 100Å pore size, 50mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-05L46S
Frequently Bought Together