ContactUsLinkComponent
C18Q x 3µm particle size x 100Å pore size, 100mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C183-03M30K
C18Q x 3µm particle size x 100Å pore size, 100mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C183-03M30K
Frequently Bought Together