ContactUsLinkComponent
C18H x 5µm particle size x 120Å pore size, 150mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Artikelnr. SGEA2C182-05N30K
C18H x 5µm particle size x 120Å pore size, 150mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Artikelnr. SGEA2C182-05N30K
Frequently Bought Together