ContactUsLinkComponent
C18H x 3µm particle size x 120Å pore size, 50mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C182-03L15K
C18H x 3µm particle size x 120Å pore size, 50mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C182-03L15K
Frequently Bought Together