Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HILIC HC x 5µm particle size x 120Å pore size, 50mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL9-05L02S
Supplier:
Trajan Scientific and Medical
Expand image
HILIC HC x 5µm particle size x 120Å pore size, 50mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL9-05L02S
Supplier:
Trajan Scientific and Medical