ContactUsLinkComponent
HILIC HC x 3µm particle size x 120Å pore size, 100mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL9-03M03S
undefined
HILIC HC x 3µm particle size x 120Å pore size, 100mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL9-03M03S
Frequently Bought Together