ContactUsLinkComponent
C18G x 3µm particle size x 200Å pore size, 50mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C181-03L30K
C18G x 3µm particle size x 200Å pore size, 50mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C181-03L30K
Frequently Bought Together