ContactUsLinkComponent
Amino x 5µm particle size x 120Å pore size, 100mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2AM-05M46S
Amino x 5µm particle size x 120Å pore size, 100mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2AM-05M46S
Frequently Bought Together