ContactUsLinkComponent
HILIC PI x 5µm particle size x 120Å pore size, 50mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL7-05L02P
undefined
HILIC PI x 5µm particle size x 120Å pore size, 50mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL7-05L02P
Frequently Bought Together