Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HILIC RP x 3µm particle size x 120Å pore size, 150mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL6-03N02P
Supplier:
Trajan Scientific and Medical
Expand image
HILIC RP x 3µm particle size x 120Å pore size, 150mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL6-03N02P
Supplier:
Trajan Scientific and Medical