Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cyano x 5µm particle size x 120Å pore size, 250mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05R03S
Supplier:
Trajan Scientific and Medical
Expand image
Cyano x 5µm particle size x 120Å pore size, 250mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CN-05R03S
Supplier:
Trajan Scientific and Medical