Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C8H x 3µm particle size x 120Å pore size, 100mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C83-03M03P
Supplier:
Trajan Scientific and Medical
Expand image
C8H x 3µm particle size x 120Å pore size, 100mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C83-03M03P
Supplier:
Trajan Scientific and Medical