Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C8H x 3µm particle size x 1000Å pore size, 150mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C82-03N46S
Supplier:
Trajan Scientific and Medical
Expand image
C8H x 3µm particle size x 1000Å pore size, 150mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C82-03N46S
Supplier:
Trajan Scientific and Medical