ContactUsLinkComponent
SCX x 3µm particle size x 120Å pore size, 50mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
  SGEA2SCX-03L02S
SCX x 3µm particle size x 120Å pore size, 50mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
  SGEA2SCX-03L02S
Frequently Bought Together