Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C8H x 3µm particle size x 1000Å pore size, 50mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C82-03L30K
Supplier:
Trajan Scientific and Medical
Expand image
C8H x 3µm particle size x 1000Å pore size, 50mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C82-03L30K
Supplier:
Trajan Scientific and Medical