Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C18G x 2.5µm particle size x 120Å pore size, 100mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-25M02P
Supplier:
Trajan Scientific and Medical
Expand image
C18G x 2.5µm particle size x 120Å pore size, 100mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-25M02P
Supplier:
Trajan Scientific and Medical