ContactUsLinkComponent
C18G x 2.5µm particle size x 120Å pore size, 50mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-25L02P
C18G x 2.5µm particle size x 120Å pore size, 50mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-25L02P
Frequently Bought Together