ContactUsLinkComponent
C18G x 5µm particle size x 120Å pore size, 150mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C185-05N15K
C18G x 5µm particle size x 120Å pore size, 150mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C185-05N15K
Frequently Bought Together