ContactUsLinkComponent
C18G x 3µm particle size x 120Å pore size, 100mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C185-03M30K
C18G x 3µm particle size x 120Å pore size, 100mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C185-03M30K
Frequently Bought Together