Spray guns
Spray gun, orange, with 6,35 mm FNPT connection
ENTE421-32-31
: ENTEGRIS
Made of high purity PFA and PTFE (all wetted parts).
- With 4,8 mm orifice and 6,35 and 12,7 mm FNPT inlet connections
- Max. pressure 5,52 bar
- Coloured caps enable visual chemical differentiation at a glance
- Description:Spray gun, orange, with 6,35 mm FNPT connection
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



