Overalls, PP
Overalls with collar, non-woven, 30 g/m², Green, XL
Catalog # HOIT129491
Supplier: HUBEI ORIENT INTL TRADING
Jump to:
Product Details & Documents
Ideal in decorating, general dust and dirt protection, factory visits, farming, food industry, general industrial handling, laboratory, painting.
- High-tech fabric that offers cloth-like comfort that is easy to wear
- Offers excellent protection against a wide range of hazards and nuisances
Weight: 22 to 55 g/m²
Weight: 22 to 55 g/m²
: Certified ISO 9001, ISO 13485, EN 340, EN 149.
Specifications
- Clothing Size:XL
- Colour:Green
- Description:Overalls with collar, non-woven, 30 g/m²
- Seq:11
- Cat. No.:HOIT129491
- Garment application:Chemical protection|Food



