Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Polypropylene-coverall white / CoverCle 100% Polypropylene, white, 55g/sqm, anti-static, CE Category 1,zipper in front, with hood 1 * 50 items
ASATCC-M
:
ASATEX
Polypropylene-coverall white / CoverCle 100% Polypropylene, white, 55g/sqm, anti-static, CE Category 1,zipper in front, with hood 1 * 50 items
ASATCC-M
:
ASATEX