Order Entry
Germany
Orders LinkContactUsLinkComponent
Overalls, PP

Overalls, PP

Overalls with hood zipper, 58 g/m², White, XXL
Catalog # HOIT120919
Overalls, PP
Catalog # HOIT120919
Overalls, PP
Overalls, PP

Overalls with hood zipper, 58 g/m², White, XXL

Price
324,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Ideal in decorating, general dust and dirt protection, factory visits, farming, food industry, general industrial handling, laboratory, painting.

  • High-tech fabric that offers cloth-like comfort that is easy to wear
  • Offers excellent protection against a wide range of hazards and nuisances

Weight: 22 to 55 g/m²

Weight: 22 to 55 g/m²

Certifications: Certified ISO 9001, ISO 13485, EN 340, EN 149.

Specifications
  • Clothing Size:
    XXL
  • Colour:
    White
  • Description:
    Overalls with hood zipper, 58 g/m²
  • Seq:
    05
  • Cat. No.:
    HOIT120919
  • Garment application:
    Chemical protection|Food