Orders LinkContactUsLinkComponent
Wafer baskets
Wafer baskets
Wafer basket, A23, 50 wafers (25 - 51 mm)
Catalog # ENTEA23-0215
Supplier:   ENTEGRIS
Wafer baskets
Catalog # ENTEA23-0215
Supplier:  ENTEGRIS
Wafer baskets
Wafer basket, A23, 50 wafers (25 - 51 mm)
Price
Inquire For Price
Log in for availability
1 * 4 ST
Product Details & Documents
These wafer baskets made of PFA are designed for processing and handling substrates. They are compatible with tanks and handles.
Compatible tanks:
A14 baskets: E12
A15 baskets: E12, E14-10-01, E22-101
A26 baskets: E14-25-01
A47 baskets: E14-21-01, E14-25-01
Compatible handles:
A14 baskets: A01, A07, A029
A15 baskets: A01, PA01, A07, PA07, A029, PA029
A26 baskets: A01, A07, A029
A47 baskets: A01, A07, A029
Specifications
  • For:
    50 wafers (25 - 51 mm)
  • Description:
    Wafer basket, A23
  • Seq:
    06
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • For A14 wafer basket
    For A14 wafer basket
    ENTEA14-02S-0215
    Log in for availability
    1 * 4 ST