Order Entry
Germany
Orders LinkContactUsLinkComponent
Beard cover, PP

Beard cover, PP

White, 240×480 mm, 100/bag
Catalog # HOIT114519
Beard cover, PP
Catalog # HOIT114519
Beard cover, PP
Beard cover, PP

White, 240×480 mm, 100/bag

Price
87,10 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

This disposable beard cover is made from soft non woven PP.

  • Single elastic headband
  • Soft, breathable and lightweight
  • Latex-free elasticated edges and headband to reduce the risk of allergic reactions

Weight: 10 to 12 g/m²

Specifications
  • Packed:
    100/bag
  • Colour:
    White
  • Seq:
    02
  • W×L:
    240×480 mm
  • Cat. No.:
    HOIT114519
  • Weight:
    10 - 12 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.