Pyronin G
Glass bottle for solids, 5 g
Catalog # CAYM14488-5
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
- CAS Number:92-32-0
- Formula:C₁₇H₁₉ClN₂O
- MDL Number:MFCD00011725
- Molecular Weight:302.8 g/mol
- Merck Index:12,08190
- Melting Pt:250…260 °C
- Storage Temperature:Ambient
- EINECS:202-147-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



