Orders LinkContactUsLinkComponent
Accessories for behrotest® compact system
Accessories for behrotest® compact system
Absorption vessel for cyanide, DIN 38405
Catalog # 717-1202
Supplier:   BEHR
Accessories for behrotest® compact system
Catalog # 717-1202
Supplier:  BEHR
Accessories for behrotest® compact system
Accessories for behrotest® compact system
Absorption vessel for cyanide, DIN 38405
Price
87,25 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Absorption vessel for cyanide
  • Type:
    DIN 38405