Order Entry
Germany
Orders LinkContactUsLinkComponent
Sampling scoops, Behrotest®

Sampling scoops, Behrotest®

Sampling scoop with borosilicate glass beaker, 1000 ml, 1 − 3,75 m, Stainless steel
Catalog # 331-3110
Supplier:   BEHR
Sampling scoops, Behrotest®
Catalog # 331-3110
Supplier:  BEHR
Sampling scoops, Behrotest®
Sampling scoops, Behrotest®

Sampling scoop with borosilicate glass beaker, 1000 ml, 1 − 3,75 m, Stainless steel

Price
441,66 €
Log in for availability
1 * 1 ST
Product Details & Documents

The sampling scoop is composed of a clamping fixture and end bar. The scoop with receptacles made of borosilicate glass comply with the high requirements.

  • Sturdy three-part supporting bar with telescopic racks
  • Continuously adjustable in length

Suitable for sampling in waters that are difficult to access, from the surface.

Certifications: EN 1485/AOX

Specifications
  • Capacity:
    1000 ml
  • Description:
    Sampling scoop with borosilicate glass beaker
  • Length:
    1 − 3,75 m
  • Material:
    Stainless steel
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.