Chromatography columns with discs
15,00 mm, 300 mm, 50 ml, 14/23 NS
WITG2.135.008
Chromatography column made of borosilicate glass with fused in sintered glass disc with ST-stopcock at the top with solid plug and screw-thread retaining nut, vacuum connection with drip tip and ST-cone at the bottom.
- Ø int.:15,00 mm
- Socket size:14/23 NS
- Capacity:50 ml
- Length:300 mm
- Seq:01



