Orders LinkContactUsLinkComponent
Inserts
E 378, insert 1/1 for a range of different utensils, with 2 carrying handles, for use in lower basket
1,7 mm mesh spacing, 5 mm frame all round, G 7882, G 7892, G 7882 CD, G 7835/36 CD, 460×460×110 mm
Catalog # MIEL3203530
Supplier:   MIELE
Inserts
Catalog # MIEL3203530
Supplier:  MIELE
Inserts
Inserts
E 378, insert 1/1 for a range of different utensils, with 2 carrying handles, for use in lower basket
1,7 mm mesh spacing, 5 mm frame all round, G 7882, G 7892, G 7882 CD, G 7835/36 CD, 460×460×110 mm
Price
427,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 378, insert 1/1 for a range of different utensils, with 2 carrying handles, for use in lower basket
    1,7 mm mesh spacing, 5 mm frame all round
  • Seq:
    11
  • W×D×H:
    460×460×110 mm
  • For:
    G 7882, G 7892, G 7882 CD, G 7835/36 CD