Order Entry
Germany
Orders LinkContactUsLinkComponent
Mounting medium, PERTEX®

Mounting medium, PERTEX®

PERTEX®, mounting medium, 1 l, 1 L
Catalog # LEIC801
Supplier:   LEICA MICROSYSTEMS
Mounting medium, PERTEX®
Catalog # LEIC801
Supplier:  LEICA MICROSYSTEMS
CAS Number:  
Mounting medium, PERTEX®

PERTEX®, mounting medium, 1 l, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents

PERTEX® is the original rapid drying medium for mounting and preserving of slide specimens. PERTEX® is proven not to affect the form, structure or colour of the mounted speci­men, even after years of storage. A thin coating of PERTEX® is enough for the cover glass to adhere to a microscope slide.

  • Xylene based medium, allows for immediate screening of slides
  • Low viscosity enable it to flow easily and prevent the formation of air bubbles
  • Neutral, colourless and has a refractive index close to that of glass at +20 °C

PERTEX® is soluble in xylene and most xylene substitutes, mounting from 99,9% ethanol is also possible.

Documents

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    PERTEX®, mounting medium, 1 l

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.