ContactUsLinkComponent
for ÄKTA start™ System High quality pre-packed columns compatible with a wide range of reducing agents and detergents. Package consisting of: 1x HiTrap Q HP Column 1 ml 1x HiTrap SP HP Column 1 ml 1x HiPrep Sephacryl S-200, 16/60 Column 5 ml 1 * 1 SET
  GEHE29-0513-25BOM
 :  Cytiva
for ÄKTA start™ System High quality pre-packed columns compatible with a wide range of reducing agents and detergents. Package consisting of: 1x HiTrap Q HP Column 1 ml 1x HiTrap SP HP Column 1 ml 1x HiPrep Sephacryl S-200, 16/60 Column 5 ml 1 * 1 SET
  GEHE29-0513-25BOM
 :  Cytiva